2-Ethoxy-3,5-bis(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

51516-500MG 51516-1G 51516-5G
77340-212EA 836.36 USD
77340-212 77352-000 77376-296
2-Ethoxy-3,5-bis(trifluoromethyl)benzoic acid ≥97%
2-Ethoxy-3,5-bis(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₈F₆O₃
MW: 302.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756803
CAS Number: 2703558-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR