Ethyl 2-(benzyloxy)-3,5-bis(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

51510-500MG 51510-1G 51510-5G
77340-364EA 940.39 USD
77340-364 77351-998 77376-294
Ethyl 2-(benzyloxy)-3,5-bis(trifluoromethyl)benzoate ≥97%
Ethyl 2-(benzyloxy)-3,5-bis(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₈H₁₄F₆O₃
MW: 392.292 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756800
CAS Number: 2764730-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR