Methyl 2-(benzyloxy)-3,5-bis(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

51509-500MG 51509-1G 51509-5G
77340-346EA 911.55 USD
77340-346 77351-996 77376-292
Methyl 2-(benzyloxy)-3,5-bis(trifluoromethyl)benzoate ≥97%
Methyl 2-(benzyloxy)-3,5-bis(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₇H₁₂F₆O₃
Storage Temperature: Ambient
MDL Number: MFCD34756799
CAS Number: 2764728-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR