Methyl 4-bromo-2,6-bis(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

51489-500MG 51489-1G 51489-5G
77335-546EA 357.41 USD
77335-546 77351-990 77376-286
Methyl 4-bromo-2,6-bis(trifluoromethyl)benzoate ≥97%
Methyl 4-bromo-2,6-bis(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₅BrF₆O₂
Storage Temperature: Ambient
MDL Number: MFCD28727930
CAS Number: 1263409-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR