2-Fluoro-4-formyl-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

51487-500MG 51487-1G 51487-5G
77338-074EA 462.47 USD
77338-074 77351-988 77376-284
2-Fluoro-4-formyl-3-(methylthio)benzoic acid ≥97%
2-Fluoro-4-formyl-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₃S
MW: 214.213 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756793
CAS Number: 2755723-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR