(4-Chloro-3-(trifluoromethoxy)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

51483-500MG 51483-1G 51483-5G 51483-25G
77328-430EA 130.81 USD
77328-430 77344-340 77364-772 77386-940
(4-Chloro-3-(trifluoromethoxy)phenyl)(methyl)sulfane ≥95%
(4-Chloro-3-(trifluoromethoxy)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₆ClF₃OS
MW: 242.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403973
CAS Number: 2386293-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR