Methyl 3-(benzyloxy)-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

51480-500MG 51480-1G 51480-5G
77328-428EA 307.97 USD
77328-428 77351-984 77376-280
Methyl 3-(benzyloxy)-5-(trifluoromethyl)benzoate ≥97%
Methyl 3-(benzyloxy)-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₃F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34564178
CAS Number: 2734778-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR