Ethyl 2-bromo-5-methoxyisonicotinate ≥97%

Supplier: AOBChem USA

Warning

51476-500MG 51476-1G 51476-5G
77335-816EA 366.68 USD
77335-816 77351-982 77376-278
Ethyl 2-bromo-5-methoxyisonicotinate ≥97%
Ethyl 2-bromo-5-methoxyisonicotinate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD28741722
CAS Number: 1276657-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR