Methyl 2,3-difluoro-6-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

51471-500MG 51471-1G 51471-5G
77336-804EA 394.49 USD
77336-804 77351-978 77376-274
Methyl 2,3-difluoro-6-(methylthio)benzoate ≥97%
Methyl 2,3-difluoro-6-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂S
MW: 218.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530120
CAS Number: 2734778-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR