2,3-Difluoro-6-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

51470-500MG 51470-1G 51470-5G 51470-25G
77328-426EA 281.19 USD
77328-426 77351-976 77364-770 77386-938
2,3-Difluoro-6-(methylthio)benzoic acid ≥97%
2,3-Difluoro-6-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂S
MW: 204.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902945
CAS Number: 2586126-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR