2-(3-Isopropyl-5-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

51467-500MG 51467-1G 51467-5G
77328-422EA 340.93 USD
77328-422 77351-972 77376-270
2-(3-Isopropyl-5-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(3-Isopropyl-5-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₅BO₂S
MW: 292.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902943
CAS Number: 2060541-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR