Ethyl 3-isopropyl-5-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

51465-500MG 51465-1G 51465-5G
77328-420EA 316.21 USD
77328-420 77351-970 77376-268
Ethyl 3-isopropyl-5-(methylthio)benzoate ≥97%
Ethyl 3-isopropyl-5-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂S
MW: 238.346 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530119
CAS Number: 2734772-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR