Methyl 3-isopropyl-5-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

51464-500MG 51464-1G 51464-5G
77328-418EA 321.36 USD
77328-418 77351-968 77376-266
Methyl 3-isopropyl-5-(methylthio)benzoate ≥97%
Methyl 3-isopropyl-5-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂S
MW: 224.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530118
CAS Number: 2734775-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR