Methyl 2-(benzyloxy)-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

51457-500MG 51457-1G 51457-5G
77336-604EA 389.34 USD
77336-604 77351-960 77376-260
Methyl 2-(benzyloxy)-5-(trifluoromethyl)benzoate ≥97%
Methyl 2-(benzyloxy)-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₃F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34530116
CAS Number: 2734778-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR