(2-Fluoro-5-iodo-3-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

51444-500MG 51444-1G 51444-5G
77339-012EA 544.87 USD
77339-012 77351-956 77376-258
(2-Fluoro-5-iodo-3-(methylthio)phenyl)methanol ≥97%
(2-Fluoro-5-iodo-3-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈FIOS
MW: 298.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178603
CAS Number: 2092373-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR