Ethyl 2-fluoro-5-iodo-3-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

51442-500MG 51442-1G 51442-5G
77337-410EA 414.06 USD
77337-410 77351-952 77376-254
Ethyl 2-fluoro-5-iodo-3-(methylthio)benzoate ≥97%
Ethyl 2-fluoro-5-iodo-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀FIO₂S
Storage Temperature: Ambient
MDL Number: MFCD34564176
CAS Number: 2734775-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR