Methyl 2-fluoro-5-iodo-3-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

51441-500MG 51441-1G 51441-5G
77336-802EA 394.49 USD
77336-802 77351-950 77376-252
Methyl 2-fluoro-5-iodo-3-(methylthio)benzoate ≥97%
Methyl 2-fluoro-5-iodo-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₉H₈FIO₂S
MW: 326.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530115
CAS Number: 2734775-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR