(2-Fluoro-5-iodo-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51438-500MG 51438-1G 51438-5G
77335-812EA 366.68 USD
77335-812 77351-948 77376-250
(2-Fluoro-5-iodo-3-methylphenyl)(methyl)sulfane ≥97%
(2-Fluoro-5-iodo-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈FIS
MW: 282.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902935
CAS Number: 2586127-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR