(2-Fluoro-3,5-diiodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51437-500MG 51437-1G 51437-5G 51437-25G
77328-406EA 202.91 USD
77328-406 77351-946 77364-760 77386-926
(2-Fluoro-3,5-diiodophenyl)(methyl)sulfane ≥97%
(2-Fluoro-3,5-diiodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅FI₂S
MW: 393.987 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28791776
CAS Number: 1806396-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR