2-Methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

51436-500MG 51436-1G 51436-5G 51436-25G
77328-404EA 212.18 USD
77328-404 77351-944 77364-758 77386-924
2-Methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2-Methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₂O
MW: 196.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11553604
CAS Number: 1256468-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR