Ethyl 2-bromo-4-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

51427-500MG 51427-1G 51427-5G
77336-076EA 375.95 USD
77336-076 77351-940 77376-246
Ethyl 2-bromo-4-isopropylbenzoate ≥97%
Ethyl 2-bromo-4-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrO₂
MW: 271.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530113
CAS Number: 2734774-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR