(2,4-Difluoro-3-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51416-1G 51416-5G 51416-25G
77341-424EA 85.49 USD
77341-424 77364-752 77376-244
(2,4-Difluoro-3-methoxyphenyl)(methyl)sulfane ≥97%
(2,4-Difluoro-3-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈F₂OS
MW: 190.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28061288
CAS Number: 1823887-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR