2-(2,4-Difluoro-3-methoxyphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

51414-1G 51414-5G 51414-25G
77341-422EA 85.49 USD
77341-422 77364-750 77376-242
2-(2,4-Difluoro-3-methoxyphenyl)propan-2-ol ≥97%
2-(2,4-Difluoro-3-methoxyphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂O₂
MW: 202.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28526447
CAS Number: 1783735-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR