1-Bromo-3-(difluoromethoxy)-5-(trifluoromethyl)benzene ≥97%
Supplier: AOBChem USA
|
Warning
|
51410-500MG
51410-1G
51410-5G
51410-25G
77328-396EA
99.91
USD
77328-396
77342-816
77364-746
77376-240
1-Bromo-3-(difluoromethoxy)-5-(trifluoromethyl)benzene ≥97%
1-Bromo-3-(difluoromethoxy)-5-(trifluoromethyl)benzene
Precaución: for Research Use Only
Formula:
C₈H₄BrF₅O MW: 291.013 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22580767 CAS Number: 1417567-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...