3-Isopropoxy-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

51407-500MG 51407-1G 51407-5G
77337-274EA 410.97 USD
77337-274 77351-936 77376-236
3-Isopropoxy-5-(trifluoromethyl)benzaldehyde ≥97%
3-Isopropoxy-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₁F₃O₂
MW: 232.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29121849
CAS Number: 1289156-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR