2,4,6-Trifluoro-N,N-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

51394-500MG 51394-1G 51394-5G 51394-25G
77328-390EA 211.15 USD
77328-390 77351-934 77364-738 77386-912
2,4,6-Trifluoro-N,N-dimethylbenzamide ≥97%
2,4,6-Trifluoro-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈F₃NO
MW: 203.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21137927
CAS Number: 1483579-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR