Morpholino(2,4,6-trifluorophenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

51391-1G 51391-5G 51391-25G
77340-704EA 60.77 USD
77340-704 77364-734 77376-232
Morpholino(2,4,6-trifluorophenyl)methanone ≥97%
Morpholino(2,4,6-trifluorophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀F₃NO₂
MW: 245.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20960342
CAS Number: 1494144-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR