6-Bromo-2-fluoro-N,N,3-trimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

51389-500MG 51389-1G 51389-5G 51389-25G
77328-388EA 126.69 USD
77328-388 77344-126 77364-730 77376-228
6-Bromo-2-fluoro-N,N,3-trimethylbenzamide ≥97%
6-Bromo-2-fluoro-N,N,3-trimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrFNO
Storage Temperature: Ambient
MDL Number: MFCD34530109
CAS Number: 2734774-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR