(6-Bromo-2-fluoro-3-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

51388-500MG 51388-1G 51388-5G 51388-25G
77328-386EA 132.87 USD
77328-386 77344-404 77364-728 77386-910
(6-Bromo-2-fluoro-3-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(6-Bromo-2-fluoro-3-methylphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrFN₂O
MW: 315.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564173
CAS Number: 2734775-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR