(6-Bromo-2-fluoro-3-methylphenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

51387-500MG 51387-1G 51387-5G 51387-25G
77328-384EA 132.87 USD
77328-384 77344-402 77364-726 77386-908
(6-Bromo-2-fluoro-3-methylphenyl)(thiomorpholino)methanone ≥97%
(6-Bromo-2-fluoro-3-methylphenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrFNOS
MW: 318.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564172
CAS Number: 2734776-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR