(6-Bromo-2-fluoro-3-methylphenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

51386-500MG 51386-1G 51386-5G 51386-25G
77328-382EA 122.57 USD
77328-382 77343-928 77364-724 77376-226
(6-Bromo-2-fluoro-3-methylphenyl)(morpholino)methanone ≥97%
(6-Bromo-2-fluoro-3-methylphenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrFNO₂
MW: 302.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530108
CAS Number: 2734779-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR