(6-Bromo-2-fluoro-3-methylphenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

51385-500MG 51385-1G 51385-5G 51385-25G
77328-380EA 126.69 USD
77328-380 77344-124 77364-722 77376-224
(6-Bromo-2-fluoro-3-methylphenyl)(piperidin-1-yl)methanone ≥97%
(6-Bromo-2-fluoro-3-methylphenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₅BrFNO
MW: 300.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530107
CAS Number: 2734774-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR