6-Bromo-N-cyclopropyl-2-fluoro-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

51383-500MG 51383-1G 51383-5G 51383-25G
77328-378EA 125.66 USD
77328-378 77344-090 77364-720 77376-222
6-Bromo-N-cyclopropyl-2-fluoro-3-methylbenzamide ≥97%
6-Bromo-N-cyclopropyl-2-fluoro-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrFNO
MW: 272.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530105
CAS Number: 1516279-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR