6-Bromo-2-fluoro-N,3-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

51382-500MG 51382-1G 51382-5G 51382-25G
77328-376EA 126.69 USD
77328-376 77344-122 77364-718 77376-220
6-Bromo-2-fluoro-N,3-dimethylbenzamide ≥97%
6-Bromo-2-fluoro-N,3-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrFNO
Storage Temperature: Ambient
MDL Number: MFCD34530104
CAS Number: 1501334-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR