6-Bromo-2-fluoro-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

51381-500MG 51381-1G 51381-5G 51381-25G
77328-374EA 87.55 USD
77328-374 77342-304 77364-716 77376-218
6-Bromo-2-fluoro-3-methylbenzamide ≥97%
6-Bromo-2-fluoro-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇BrFNO
Storage Temperature: Ambient
MDL Number: MFCD34530103
CAS Number: 1532714-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR