(2-Fluoro-3-methoxy-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51377-500MG 51377-1G 51377-5G 51377-25G
77328-372EA 225.57 USD
77328-372 77351-932 77364-714 77386-906
(2-Fluoro-3-methoxy-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(2-Fluoro-3-methoxy-6-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₈F₄OS
MW: 240.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530101
CAS Number: 2705984-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR