2-Fluoro-1-methoxy-3-methyl-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

51375-500MG 51375-1G 51375-5G 51375-25G
77328-368EA 130.81 USD
77328-368 77344-338 77364-710 77386-902
2-Fluoro-1-methoxy-3-methyl-4-(trifluoromethyl)benzene ≥95%
2-Fluoro-1-methoxy-3-methyl-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈F₄O
MW: 208.153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564171
CAS Number: 2385078-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR