Methyl 2-fluoro-3-methoxy-6-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2-Fluoro-3-methoxy-6-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

51373-500MG 51373-1G 51373-5G 51373-25G
77328-364EA 146.26 USD
77328-364 77345-164 77364-706 77386-898
Methyl 2-fluoro-3-methoxy-6-(trifluoromethyl)benzoate ≥97%
Methyl 2-fluoro-3-methoxy-6-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₃
MW: 252.162 g/mol
Storage Temperature: Ambient
CAS Number: 2703027-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR