2-Fluoro-3-methoxy-6-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

51372-500MG 51372-1G 51372-5G 51372-25G
77328-362EA 120.51 USD
77328-362 77343-738 77364-704 77376-216
2-Fluoro-3-methoxy-6-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-3-methoxy-6-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₃
MW: 238.136 g/mol
Storage Temperature: Ambient
CAS Number: 2386363-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR