1-(Difluoromethoxy)-2-fluoro-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

51366-500MG 51366-1G 51366-5G
77340-256EA 862.11 USD
77340-256 77351-930 77376-214
1-(Difluoromethoxy)-2-fluoro-4-(trifluoromethyl)benzene ≥97%
1-(Difluoromethoxy)-2-fluoro-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₄F₆O
MW: 230.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28789467
CAS Number: 1803778-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR