1-(Benzyloxy)-2-fluoro-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

51365-500MG 51365-1G 51365-5G
77328-360EA 332.69 USD
77328-360 77351-928 77376-212
1-(Benzyloxy)-2-fluoro-4-(trifluoromethyl)benzene ≥97%
1-(Benzyloxy)-2-fluoro-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₄O
MW: 270.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530095
CAS Number: 1044067-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR