2-Fluoro-1-(methoxymethoxy)-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

51364-500MG 51364-1G 51364-5G
77335-750EA 364.62 USD
77335-750 77351-926 77376-210
2-Fluoro-1-(methoxymethoxy)-4-(trifluoromethyl)benzene ≥95%
2-Fluoro-1-(methoxymethoxy)-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈F₄O₂
MW: 224.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530094
CAS Number: 2734778-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR