2-Chloro-1-iodo-5-methoxy-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

51363-500MG 51363-1G 51363-5G 51363-25G
77328-358EA 226.6 USD
77328-358 77351-924 77364-702 77386-896
2-Chloro-1-iodo-5-methoxy-3-(trifluoromethyl)benzene ≥97%
2-Chloro-1-iodo-5-methoxy-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₅ClF₃IO
MW: 336.477 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756786
CAS Number: 2386202-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR