(2-Chloro-5-methoxy-3-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

51362-500MG 51362-1G 51362-5G
77336-800EA 394.49 USD
77336-800 77351-922 77376-208
(2-Chloro-5-methoxy-3-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(2-Chloro-5-methoxy-3-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₈ClF₃OS
MW: 256.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756785
CAS Number: 2706393-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR