2-Chloro-5-methoxy-3-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

51357-500MG 51357-1G 51357-5G
77336-908EA 399.64 USD
77336-908 77351-920 77376-206
2-Chloro-5-methoxy-3-(trifluoromethyl)benzoic acid ≥97%
2-Chloro-5-methoxy-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₃
MW: 254.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756780
CAS Number: 2387276-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR