(3-Ethoxy-5-(trifluoromethyl)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

51351-500MG 51351-1G 51351-5G
77337-950EA 454.23 USD
77337-950 77351-918 77376-202
(3-Ethoxy-5-(trifluoromethyl)phenyl)methanol ≥97%
(3-Ethoxy-5-(trifluoromethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD32633289
CAS Number: 2387134-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR