4-Chloro-2-(difluoromethoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

51343-1G 51343-5G 51343-25G
77340-900EA 62.83 USD
77340-900 77364-698 77376-196
4-Chloro-2-(difluoromethoxy)benzoic acid ≥95%
4-Chloro-2-(difluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₅ClF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD12828089
CAS Number: 97914-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR