2-Chloro-4-fluoro-3-iodo-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

51333-1G 51333-5G 51333-25G
77342-012EA 111.24 USD
77342-012 77364-694 77376-192
2-Chloro-4-fluoro-3-iodo-1-methylbenzene ≥97%
2-Chloro-4-fluoro-3-iodo-1-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFI
MW: 270.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751657
CAS Number: 1502669-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR