1,3-Dichloro-2-iodo-4-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

51328-500MG 51328-1G 51328-5G 51328-25G
77328-354EA 99.91 USD
77328-354 77342-814 77364-692 77376-190
1,3-Dichloro-2-iodo-4-methoxybenzene ≥97%
1,3-Dichloro-2-iodo-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂IO
MW: 302.924 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178600
CAS Number: 2384443-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR