1,3-Dichloro-4-methoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

51327-500MG 51327-1G 51327-5G 51327-25G
77328-352EA 187.46 USD
77328-352 77351-908 77364-690 77386-892
1,3-Dichloro-4-methoxy-2-methylbenzene ≥97%
1,3-Dichloro-4-methoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O
MW: 191.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28360087
CAS Number: 1803824-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR